1-Octylphosphonic dichloride 97%

Supplier: Thermo Fisher Scientific
Danger

L18587.14 L18587.06
L18587.14EA 894 EUR
L18587.14 L18587.06
1-Octylphosphonic dichloride 97%
1-Octylphosphonic dichloride

Purity: 97% CAS No.: 3095-94-1 Molecular Formula: C8H17Cl2OP Formula Weight: 231.10 Refractive Index: 1.4665 Boiling Point: 143-145°/12mm Beilstein Reference: 1931202 UN No.: UN3265 MDL No.: MFCD00015813

Formula: C₈H₁₇Cl₂OP
Boiling Pt: 143…145 °C (12 mmHg)
Density: 1.115
Storage Temperature: Ambient
MDL Number: MFCD00015813
CAS Number: 3095-94-1
EINECS: 221-444-3
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR