Safety cabinets for gas cylinders, SiZ G30
Supplier: LACONT UMWELTTECHNIK
B80-5017-C B80-5015-C B80-5016-C B80-5018-C
LACOB80-5017-CEA 0 EUR
LACOB80-5017-C LACOB80-5015-C LACOB80-5016-C LACOB80-5018-C
Safety cabinets for gas cylinders, SiZ G30
Furniture Storage Cabinets
Safety cabinets are approved for storage of pressurised gas cylinders at the workplace.
  • Stainless steel base plate- Niro Rhombus
  • Leveling feet for installing cabinet horizontally
  • Doors with cylinder lock
  • Automatic thermal closing mechanism if fire occurs
Outer body made from powder-coated sheet steel, doors either in RAL 7035 - light grey or RAL 1007 - daffodil yellow and inner body made from HPL-coated sheets - RAL 7035.
Certifications: Conforms with DIN EN 14470-2 (G 30), DIN EN 14470-2, GS-approved safety (GS-sign), DIN 4102.