Chemical cabinets for small containers
Supplier: LACONT UMWELTTECHNIK
B80-6215-A B80-6207-A B80-6208-A B80-6214-A
LACOB80-6215-AEA 0 EUR
LACOB80-6215-A LACOB80-6207-A LACOB80-6208-A LACOB80-6214-A
Chemical cabinets for small containers
Furniture Storage Cabinets
Chemical and poison cabinets are ideal for safe and regular storage of non-flammable hazardous substances, chemicals and flammable liquids in workspaces.
  • Doors with central cylinder lock and ventilation slots
  • Central DN 80 ventilation connection for exhaust air extraction of both cabinets on roof
  • Available with six door colours
  • Automatic thermal closing mechanism in case of fire
Cabinets are made of single-wall body made from high-quality powder-coated sheet steel in RAL 7035 - light grey Model GL with acrylic glass doors. It also contains smallMore Product Information