Condenser with cooling coil and jacket, with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05250214 05250314 05250414 05250211 05250313 05250514
201-1681EA 122 EUR
201-1681 201-1682 201-1683 LENZ05250211 LENZ05250313 LENZ05250514
Condenser with cooling coil and jacket, with standard ground joints
Condensers
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Additional cooling surface for condensing highly volatile vapours. Cooling coil and cooling jacket, with standard ground cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
DIN 12593
Certifications: DIN 12593