Adapters, delivery, straight, with drain tip
Supplier: Lenz Laborglas GmbH & CO.KG
05266003 05266002 05266001 05266004
LENZ05266003EA 24.9 EUR
LENZ05266003 LENZ05266002 LENZ05266001 LENZ05266004
Adapters, delivery, straight, with drain tip
Distillation Components Distillation Adapters
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Delivery adapters, straight, with NS socket and NS cone, with drain tip.