Distilling receivers, Anschütz-Thiele, straight, with cooling jacket
Supplier: Lenz Laborglas GmbH & CO.KG
05305004 05305014 05305104 05305114
LENZ05305004EA 656 EUR
LENZ05305004 LENZ05305014 LENZ05305104 LENZ05305114
Distilling receivers, Anschütz-Thiele, straight, with cooling jacket
Distillation Components Distillation Receivers
Made of DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Receivers with straight adapter, with cooling jacket, graduated, with socket and cone NS 29/32, vacuum connection: thread GL18, NS stopcock with solid glass or PTFE plug.