Perfection and Performance Stainless-Steel Refrigerators for Professional Use, FRPSvh & FRPSvg
Supplier: Liebherr
FRPSVH6501 FRPSVG6501 FRPSVH6511 FRPSVG6511 FRPSVH1402 FRPSVG1402 FRPSVH1412
LIEBFRPSVH6501EA 3999 EUR
LIEBFRPSVH6501 LIEBFRPSVG6501 LIEBFRPSVH6511 LIEBFRPSVG6511 LIEBFRPSVH1402 LIEBFRPSVG1402 LIEBFRPSVH1412
Perfection and Performance Stainless-Steel Refrigerators for Professional Use, FRPSvh & FRPSvg
Refrigerators
Glass/steel door high-volume, general-purpose refrigerators with forced air cooling. Stainless-steel exterior and interior. Temperature stability in accordance with EN 16825.
  • Energy-efficiency optimised appliances for lower running costs and help to meet user sustainability targets
  • Equipped with 150 mm legs, allows cleaning underneath
  • Mechanical door locks for basic content security
  • Forced-air cooling system with energy-efficient hot gas defrost
  • Power failure alarm when mains power returns
Choose between two execution options to suit differingMore Product Information