TLC adsorbents, Polyamide 6
Supplier: MACHEREY-NAGEL
816610.1 816620.1
MANA816610.1EA 735 EUR
MANA816610.1 MANA816620.1
TLC adsorbents, Polyamide 6
Chromatography Sorbents TLC Sorbents
Polyamide 6 adsorbents for thin layer chromatography.
  • High-quality polyamide
  • Special separation mechanism (hydrogen bonds to amide groups)
  • Especially recommended for natural compounds
  • Available with or without fluorescence indicator