Cavity slides for diagnosis
Supplier: MARIENFELD
1216550 1216751 1216521 1216542 1215130 1216690 1216650 1215671 1216750 1216820 1216492 1215672 1216652 1215670 1216330 1216680 1215132 1216530 1216532 1216540 1216682 1216071 1216334 1216824 1215131 1216331 1216491 1216681 1216821 1216520 1216523 1216691 1216541 1216551 1216651 1216531
MARI1216550EA 83.6 EUR
MARI1216550 MARI1216751 MARI1216521 MARI1216542 MARI1215130 MARI1216690 MARI1216650 MARI1215671 MARI1216750 MARI1216820 MARI1216492 MARI1215672 MARI1216652 MARI1215670 MARI1216330 MARI1216680 MARI1215132 MARI1216530 MARI1216532 MARI1216540 MARI1216682 MARI1216071 MARI1216334 MARI1216824 MARI1215131 MARI1216331 MARI1216491 MARI1216681 MARI1216821 MARI1216520 MARI1216523 MARI1216691 MARI1216541 MARI1216551 MARI1216651 MARI1216531
Cavity slides for diagnosis
Slides Cavity Slides
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
  • Soda-lime glass with epoxy resin colour masks
  • High resistance against solvents