Ceramic Blade No. 92C

Supplier: MARTOR ARGENTAX
1792.25
MARO1792.25EA 20.5 EUR
MARO1792.25
Ceramic Blade No. 92C
Razors and Blades
Ceramic blade no. 92C is a very high-quality blade made of ceramic.

  • Ceramic
  • Non-magnetic
  • Non-conductive
  • Chemically inert
  • Non-oiled
  • Non-rusting

It remains very sharp for a very long time and has four cutting edges that can be used one after the other for an extra long service life. The wide ground edge makes cutting easier. The blade is 26 mm long, 18,5 mm wide and 0,50 mm thick.

Refinement/version: Stainless and acid resistant
Ground: Deep-edged, two sided ground-edge
Blade facets: Double bevel edge-ground
Blade characteristics: Four cutting edgeMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR