Culture media in bottles, CHROMagar™
Supplier: CHROMAGAR
MR502 SB282 ESRT2 15EC168
MR502EA 501 EUR
MR502 SB282 ESRT2 MAST15EC168
Culture media in bottles, CHROMagar™
Media for Microbiology Prepared Media for Microbiology
With traditional media identification of mixed cultures is difficult, whereas CHROMagar™ just by the colour of the colony will instantly differentiate various species.
  • High contrast
  • Fast
  • Convenient