N-Methyl-4-nitroaniline

Supplier: Thermo Scientific

SB01115.1 SB01115.10 SB01115.50
MAYBSB01115.1EA 0 EUR
MAYBSB01115.1 MAYBSB01115.10 MAYBSB01115.50
N-Methyl-4-nitroaniline
N-Methyl-4-nitroaniline
Formula: C₇H₈N₂O₂
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 148…149 °C
Density: 1.201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007305
CAS Number: 100-15-2
EINECS: 202-823-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR