4'-Nitroacetophenone

Supplier: Thermo Scientific

Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone

SB01771.1 SB01771.10 SB01771.50
MAYBSB01771.1EA 0 EUR
MAYBSB01771.1 MAYBSB01771.10 MAYBSB01771.50
4'-Nitroacetophenone
4'-Nitroacetophenone
Formula: C₈H₇NO₃
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 76…78 °C
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007355
CAS Number: 100-19-6
EINECS: 202-827-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR