High-density shelving system, MetroMax i™ Top-Track™
Supplier: Metro International
MXTTE24 MXTTM18C MXTTM24C MXTTE18 MXTTM18S
METOMXTTE24EA 0 EUR
METOMXTTE24 METOMXTTM18C METOMXTTM24C METOMXTTE18 METOMXTTM18S
High-density shelving system, MetroMax i™ Top-Track™
Furniture Shelves
MetroMax i™ Top-Track™ shelving system maximises the usable storage area in a given space.
  • Maximum load rating for a single unit is 408,2 kg
  • Corrosion proof provides solid shelf option and better chemical resistance
  • All-polymer construction helps for high-moisture applications
  • Chrome-plated units are ideal for dry environments with minimal exposure to corrosives
  • Stainless steel units provide maximum protection in corrosive environments
A movable, open aisle allows full access to each entire shelving unit helps to eliminating hard to reach or deadMore Product Information