Accessories for Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Supplier: Avantor Fluid Handling
06043-80
MFLX06043-80EA 132 EUR
MFLX06043-80
Accessories for Bürkle Stainless Steel Barrel Pumps for Flammable Liquids
Pumps Drum/Transfer Pumps
Compatible with the Bürkle Stainless Steel Barrel Pumps For flammable Liquids.
  • Safely transfer flammable and combustible liquids with ease
  • Constructed of conductive 304 Stainless Steel for enhanced durability and safety
Safely transfer flammable liquids with the stainless steel barrel pumps.
There are two pump models to choose from. The stainless steel barrel pumps with discharge tube feature a spout-type outlet. The drum pumps with discharge tube and stopcock are suitable for fluid transfer in which the outlet container is further from the fillingMore Product Information