CPC® Quick-Disconnect Fittings, Compression Inserts
Supplier: Avantor Fluid Handling
06360-77 06361-32 06361-32BLK 06360-77BLK 31307-54BLK 31307-44BLK
MFLX06360-30EA 13.3 EUR
MFLX06360-30 MFLX06360-41 MFLX06360-74 MFLX06360-75 MFLX06364-30 MFLX06364-75 MFLX06360-77 MFLX06361-32 MFLX06361-32BLK MFLX06360-77BLK MFLX06360-74BLK MFLX06364-30BLK MFLX06360-41BLK MFLX06360-30BLK MFLX06360-75BLK MFLX06364-75BLK MFLX31307-54BLK MFLX31307-44BLK
CPC® Quick-Disconnect Fittings, Compression Inserts
Fittings and Connectors
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information