Masterflex® I/P® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Supplier: Avantor Fluid Handling
MFLX06401-73EA 579 EUR
MFLX06401-73 MFLX06401-82 MFLX06401-26
Masterflex® I/P® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
Designed to resist the swelling and embrittlement caused by hydrocarbon-based fluids.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
Masterflex® I/P® Tygon® Fuel and Lubricant Precision pump tubing is speciallyMore Product Information