CPC® High-Flow Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: Avantor Fluid Handling
31313-13BLK
MFLX31313-11BLKEA 134 EUR
MFLX31313-11BLK MFLX31313-13BLK
CPC® High-Flow Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Fittings and Connectors
Fittings connect in a snap.
  • Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
  • Fitting body constructed of durable polysulfone
These CPC® quick-disconnect fittings provide aseptic disconnect functionality. The thumb latch enables easy one-handed operation. An audible click is heard when the body and insert are coupled to confirm a secure, leak-free connection. These fittings are made of USP Class VI-compliant and BSE/TSE-free polysulfone, ideal for rugged applications that require material certification.
For aMore Product Information