Masterflex® 1-1/2" Sanitary Clamp High Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLX40259-23EA 107 EUR
MFLX40259-23 MFLX40259-24 MFLX40259-25 MFLX40259-26 MFLX40259-27 MFLX40259-28 MFLX40259-29 MFLX40259-30 MFLX40259-31 MFLX40259-32 MFLX40259-33 MFLX40259-34 MFLX40259-35 MFLX40259-36 MFLX40259-37 MFLX40259-38 MFLX40259-39 MFLX40259-40
Masterflex® 1-1/2" Sanitary Clamp High Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information