Masterflex® Stationary Male Luer Lock to Barb Fitting, Avantor®
Supplier: Avantor Fluid Handling
MFLX40322-00EA 16.2 EUR
MFLX40322-00 MFLX40322-01 MFLX40322-02 MFLX40322-03 MFLX40322-04 MFLX40322-05 MFLX40322-06 MFLX40322-07 MFLX40322-08 MFLX40322-09 MFLX40322-10 MFLX40322-11 MFLX40322-12 MFLX40322-13 MFLX40322-14 MFLX40322-15 MFLX40322-16 MFLX40322-17
Masterflex® Stationary Male Luer Lock to Barb Fitting, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information