Masterflex® ¾” Sanitary Tri-Clamp to Hose Barb Adapter, Avantor®
Supplier: Avantor Fluid Handling
MFLX40330-03EA 274 EUR
MFLX40330-03 MFLX40330-04 MFLX40330-05 MFLX40330-06 MFLX40330-07 MFLX40330-08 MFLX40330-09 MFLX40330-10 MFLX40330-11 MFLX40330-12 MFLX40330-13 MFLX40330-14 MFLX40330-15 MFLX40330-16 MFLX40330-17 MFLX40330-18
Masterflex® ¾” Sanitary Tri-Clamp to Hose Barb Adapter, Avantor®
Fittings and Connectors
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in theMore Product Information