Masterflex® Drum Pump Replacement Motors, Avantor®
Supplier: Avantor Fluid Handling
70604-03 70604-13 70604-16
MFLX70604-03EA 0 EUR
MFLX70604-03 MFLX70604-13 MFLX70604-16
Masterflex® Drum Pump Replacement Motors, Avantor®
Pumps Motors
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Use with Masterflex® Complete Drum and Container Pumps
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks and small storage vessels. These Masterflex® drum pump replacement motors are one component of a system that includes a pump tube and motor (order pump tubes separately). Motors are equipped with variable-speed control for added safety and control.