Masterflex® B/T® MasterSense™ BioPharma Process Pump, Avantor®
Supplier: Avantor Fluid Handling
MFLX77112-10-EUEA 15970 EUR
MFLX77112-10-EU MFLX77112-10-UK MFLX77112-10-IT
Masterflex® B/T® MasterSense™ BioPharma Process Pump, Avantor®
Pumps Peristaltic Pumps
High-flow pump for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful pump engineered for continuous-duty biopharma process applications
  • 316 stainless steel housing is IP66 rated for use in washdown environments
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Adjust parameters with a simple touch, even while wearing gloves
  • Over-the-air firmware updates let you take full advantage of future improvements in pump features and functionality
  • Options for connectivity and