Masterflex® L/S® MasterSense™ Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
MFLX77920-42-EUEA 5750 EUR
MFLX77920-42-EU MFLX77920-42-UK MFLX77920-42-IT MFLX77976-00-EU MFLX77976-02-EU MFLX77976-10-EU MFLX77976-12-EU MFLX77920-50-EU MFLX77976-00-UK MFLX77976-02-UK MFLX77976-10-UK MFLX77976-12-UK MFLX77920-50-UK MFLX77976-00-IT MFLX77976-02-IT MFLX77976-10-IT MFLX77976-12-IT MFLX77920-50-IT MFLX77920-40-EU MFLX77920-52-EU MFLX77920-40-UK MFLX77920-52-UK MFLX77920-40-IT MFLX77920-52-IT
Masterflex® L/S® MasterSense™ Pump Systems, Avantor®
Pumps Peristaltic Pumps
Complete pump systems for accurate fluid delivery - with an intuitive touch-screen interface.
  • Precise, repeatable performance that is quick and easy to set up and program
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include MasterflexLive, EtherNet/IP™, Profibus, or analog signal