Masterflex® I/P® MasterSense™ pump systems
Supplier: Avantor Fluid Handling
MFLX77966-10EA 12030 EUR
MFLX77966-10 MFLX77966-00 MFLX77966-00-EU MFLX77966-10-EU MFLX77966-00-UK MFLX77966-10-UK MFLX77966-00-IT MFLX77966-10-IT MFLX77966-00-IL MFLX77966-10-IL
Masterflex® I/P® MasterSense™ pump systems
Pumps Peristaltic Pumps
Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful, space-efficient drives will accept all I/P pump head and tubing options
  • 17,8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include EtherNet/IP™, Profibus, analog signal