Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
MFLX78006-22EA 6930 EUR
MFLX78006-22 MFLX78006-24 MFLX78006-26 MFLX78006-28 MFLX78006-42 MFLX78006-44 MFLX78006-46 MFLX78006-48 MFLX78006-22-AU MFLX78006-24-AU MFLX78006-26-AU MFLX78006-28-AU MFLX78006-42-AU MFLX78006-44-AU MFLX78006-46-AU MFLX78006-48-AU MFLX78006-22-DK MFLX78006-24-DK MFLX78006-26-DK MFLX78006-28-DK MFLX78006-42-DK MFLX78006-44-DK MFLX78006-46-DK MFLX78006-48-DK MFLX78006-22-IN MFLX78006-24-IN MFLX78006-26-IN MFLX78006-28-IN MFLX78006-42-IN MFLX78006-44-IN MFLX78006-46-IN MFLX78006-48-IN MFLX78006-22-EU MFLX78006-24-EU MFLX78006-26-EU MFLX78006-28-EU MFLX78006-42-EU MFLX78006-44-EU MFLX78006-46-EU MFLX78006-48-EU MFLX78006-22-UK MFLX78006-24-UK MFLX78006-26-UK MFLX78006-28-UK MFLX78006-42-UK MFLX78006-44-UK MFLX78006-46-UK MFLX78006-48-UK MFLX78006-22-CH MFLX78006-24-CH MFLX78006-26-CH MFLX78006-28-CH MFLX78006-42-CH MFLX78006-44-CH MFLX78006-46-CH MFLX78006-48-CH MFLX78006-22-IT MFLX78006-24-IT MFLX78006-26-IT MFLX78006-28-IT MFLX78006-42-IT MFLX78006-44-IT MFLX78006-46-IT MFLX78006-48-IT MFLX78006-22-IL MFLX78006-24-IL MFLX78006-26-IL MFLX78006-28-IL MFLX78006-42-IL MFLX78006-44-IL MFLX78006-46-IL MFLX78006-48-IL
Masterflex® Ismatec® IPC High-Accuracy Multichannel Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Achieve high-accuracy multi-channel flow, with an intuitive touch-screen interface.
  • Replicate same flow for all channels; perfect for auto-analysis, reactor studies, cell line seeding, and research
  • Planetary drive with 8 stainless steel rollers reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • 12,7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or remote control (analog remote control, serial, or EtherNet/IP)
  • Three-level user access – gives you the