Masterflex® Ismatec® IP and IP-N Digital Multichannel Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
78010-03
MFLX78010-09-EUEA 5070 EUR
MFLX78010-09-EU MFLX78010-27-EU MFLX78010-29-EU MFLX78010-47-EU MFLX78011-09-EU MFLX78011-17-EU MFLX78011-29-EU MFLX78011-39-EU MFLX78011-47-EU MFLX78010-09-UK MFLX78010-27-UK MFLX78010-29-UK MFLX78010-47-UK MFLX78011-09-UK MFLX78011-17-UK MFLX78011-29-UK MFLX78011-39-UK MFLX78011-47-UK MFLX78010-09-IT MFLX78010-27-IT MFLX78010-29-IT MFLX78010-47-IT MFLX78011-09-IT MFLX78011-17-IT MFLX78011-29-IT MFLX78011-39-IT MFLX78011-47-IT MFLX78010-03 MFLX78010-39-EU MFLX78010-49-EU MFLX78010-59-EU MFLX78011-07-EU MFLX78011-27-EU MFLX78010-39-UK MFLX78010-49-UK MFLX78010-59-UK MFLX78011-07-UK MFLX78011-27-UK MFLX78010-39-IT MFLX78010-49-IT MFLX78010-59-IT MFLX78011-07-IT MFLX78011-27-IT
Masterflex® Ismatec® IP and IP-N Digital Multichannel Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Smooth, precise flow and fewer tubing changes.
  • Planetary drive with 8 stainless steel rotors reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • Four-digit LED display facilitates repeatability
  • Control pump via 6-button keypad or analog and TTL external signals
  • Automatic occlusion cartridges are easy to use and provide reproducible results
These Ismatec® IP and IP-N digital peristaltic multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. TheMore Product Information