Masterflex® L/S® Spooled Pump Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
MFLX95687-14EA 2700 EUR
MFLX95687-14 MFLX95687-15 MFLX95687-16 MFLX95687-17 MFLX95687-18 MFLX95687-24 MFLX95687-35 MFLX95687-36
Masterflex® L/S® Spooled Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly