Masterflex® Ismatec® Pump Tubing with Flared Ends, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Supplier: Avantor Fluid Handling
MFLX97617-10EA 67.4 EUR
MFLX97617-10 MFLX97617-12 MFLX97617-14 MFLX97617-18 MFLX97617-26 MFLX97617-28
Masterflex® Ismatec® Pump Tubing with Flared Ends, 3-Stop Microbore, Puri-Clear™ LL, Avantor®
Tubing Peristaltic Pump Tubing
Flared ends for easy extension tubing assembly.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubingMore Product Information