Laboratory Glassware Washers, Large Capacity, PLW 8616
Supplier: MIELE
11046430 11046490 11046440 11046510 10928990 10931290 10931330 10941940 10880710 11332020
MIEL11046430EA 54680 EUR
MIEL11046430 MIEL11046490 MIEL11046440 MIEL11046510 MIEL10928990 MIEL10931290 MIEL10931330 MIEL10941940 MIEL10880710 MIEL11332020
Laboratory Glassware Washers, Large Capacity, PLW 8616
Glassware Washing Machines
The PLW 8616 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to use, it has modular load carriers and SimpleLoad system. This efficient unit has a double door, 900 mm wide, a usable capacity of 351 litres. PLW 8616 provides reliable results, high pump performance with a variable-speed pump and features spray arm monitoring and a conductivity meter.
  • Innovative 2-pump system for maximum performance
  • New flexible and modular (SimpleLoad) racking system
  • Excellent drying