Prefilters and Net Filters
Supplier: Merck
AN3H04700 AN0604700 AN1204700 AN2504700 PP2514250 PP4502500 PP4504700 PP4509030 PP2504700 PP2502500 AN1H04700 AN5004700
MILFAN3H04700EA 198 EUR
MILFAN3H04700 MILFAN0604700 MILFAN1204700 MILFAN2504700 MILFPP2514250 MILFPP4502500 MILFPP4504700 MILFPP4509030 MILFPP2504700 MILFPP2502500 MILFAN1H04700 MILFAN5004700
Prefilters and Net Filters
Filters Membrane Filters
These PP white filters are designed for general clarification and prefiltration of solvents, deionized water and bioburden reduction.
  • Broad thermal compatibility
  • Compatible with aggressive solvents
The prefilters provide high particle retention, high dirt-holding capacity and low pressure drop. They are designed for filtration of organic solvents, but they can also be used, once properly wetted with alcohol, for the filtration of aqueous solutions. They are ideal for general prefiltration and clarification, suitable for organic solvents.