Solvent Filtering Dispenser

Supplier: Merck
XX6602500
MILFXX6602500EA 768 EUR
MILFXX6602500
Solvent Filtering Dispenser
Bottle Top Dispensers
Ultraclean and dispense small volumes of solvent by squeeze-bottle action. This hand-pressure operated dispenser features a filter holder fitted with a delivery tube. Use to direct filtered solvent against surfaces.

  • Integrated hand pump eliminates the need for external pump
  • Holds up to 1 L of solvent

Applications: Solvent filtration prior to contamination analysis, solvent rinsing of machined parts and collection containers.

Components: TFE gasket, PTFE seal, PTFE tubing, borosilicate glass flask, glass elbow(s), glass nozzle (for dispensing),More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR