SpectraMax ABS Plus UV/Vis Single-Mode Microplate Reader with built-in Cuvette Port. System Includes: Tunable Monochromator-based Sp ectral Absorbance (190nm to 1000nm), SoftMax Pro Software (4 perpetual licenses) Integrated Linear Shaking and Tempe 1 * 1 items
Supplier: Molecular Devices
SpectraMax ABS Plus UV/Vis Single-Mode Microplate Reader with built-in Cuvette Port. System Includes: Tunable Monochromator-based Sp ectral Absorbance (190nm to 1000nm), SoftMax Pro Software (4 perpetual licenses) Integrated Linear Shaking and Tempe 1 * 1 items