Microplate reader, Gemini™ EM
Supplier: Molecular Devices
EM
MLDVEMEA 0 EUR
MLDVEM
Microplate reader, Gemini™ EM
Microplate Readers
Gemini™ EM microplate spectrofluorometer provides a flexible environment to determine the optimal excitation and emission settings for most fluorescent intensity assays. The Gemini EM reader with dual monochromators allows researchers to try new and novel dyes without having to purchase expensive filter sets.
  • Measure fluorescence from solution- or cell-based assays by switching between top and bottom fluorescence reading
  • Up to 4 wavelength pairs can be read for endpoint and kinetic measurements
  • Read a variety of sample formats from 6- to 384-well