Aura™ Health Care Particulate Respirators 1861+, 1862+ and 1863+
Supplier: 3M
1861PLUS 1862PLUS
MMMA1861PLUSEA 38.2 EUR
MMMA1861PLUS MMMA1862PLUS
Aura™ Health Care Particulate Respirators 1861+, 1862+ and 1863+
Respirators Disposable Respirators Particulate Respirators
The 3M™ Aura™ Health Care Particulate Respirators provide effective respiratory protection for use in medical environments where health care workers will be exposed to airborne dust particles, non-volatile liquid particles and bioaerosols. These respirators limit the transmission of infective agents from staff to patients and are suitable for use during surgical procedures and certain other medical procedures. These products also offer resistance to penetration of splashes of liquid.
  • Sculpted nose panel helps conforms to the nose and contours of the face