Earplugs, Tri-Flange™
Supplier: 3M
PN01006
MMMAPN01006EA 1030 EUR
MMMAPN01006
Earplugs, Tri-Flange™
Earplugs Reusable Earplugs
3M™ Tri-Flange™ Earplugs are innovative, comfortable hearing protectors with soft, flexible flanges that adapt to the shape of ears.
  • Reduces noise levels up to 29 dB
  • Features soft, flexible triple flanges that adapt to the shape of the ear for an effective acoustic seal
  • No need to handle the plug tip, simply push the plug to fit
  • Ideal for use in dirty environments
  • Easy to wash and reuse
  • Cloth cord
Get comfortable and hygienic hearing protection with 3M™ Tri-Flange™ Earplugs. Reliable and robust, the plugs are moisture-resistant making them suitableMore Product Information