Biological Indicator Incubator, 116, Attest™
Supplier: 3M MEDICAL
M118
MMMEM118EA 1000 EUR
MMMEM118
Biological Indicator Incubator, 116, Attest™
Incubators Standard Incubators
3M™ Attest™ incubators provide optimal conditions for accurate readout using biological indicators.
  • Nominal voltage required is 240 V
  • 15-well capacity
  • Dry block pre-set to 56 °C incubation temperature
  • Manage record-keeping with web app and your internet connected PC
The incubator's internal temperatures are rigidly controlled to 56 ±2 °C (133 ±3 °F) and automatically maintained. If a temperature measurement device is used to monitor the incubator, the sensor should be in contact with the metal heating block wall.