Waterproof digital thermometer, K-type
Supplier: MOINEAU
3134T
MOIN3134TEA 290 EUR
MOIN3134T
Waterproof digital thermometer, K-type
Thermometers Digital Thermometers
Digital thermometer with K-type thermocouple probe. Waterproof and therefore washable: Ideal for food and kitchen applications.
  • Magnet option: thermometer can be attached to any surface
  • Auto power-off after 10 minutes
  • 'HOLD' function
Delivery information: Supplied with wired glass silk +400 °C probe and CR2032 lithium battery.