Cycloheximide

Supplier: Molekula
Danger

Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,

15065373-5G
MOLE15065373-5GEA 394 EUR
MOLE15065373-5G
Cycloheximide
Cycloheximide
Formula: C₁₅H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR