Ammonium thiocyanate

Supplier: Molekula
Warning

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

18466896-1KG
MOLE18466896-1KGEA 71 EUR
MOLE18466896-1KG
Ammonium thiocyanate
Ammonium thiocyanate
Formula: NH₄SCN
Melting Pt: 150 °C
Density: 1.305 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR