Sodium pyruvate

Supplier: Molekula

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

19010543-500G 19010543-100G
MOLE19010543-500GEA 250 EUR
MOLE19010543-500G MOLE19010543-100G
Sodium pyruvate
Sodium pyruvate
Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR