Sodium borohydride

Supplier: Molekula
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

19526770-100G
MOLE19526770-100GEA 250 EUR
MOLE19526770-100G
Sodium borohydride
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR