(1-Butyl)triphenylphosphonium bromide

Supplier: Molekula
Warning

20583883-100G
MOLE20583883-100GEA 81.8 EUR
MOLE20583883-100G
(1-Butyl)triphenylphosphonium bromide
(1-Butyl)triphenylphosphonium bromide
Formula: H₃C(CH₂)₃P(C₆H₅)₃Br
Melting Pt: 249…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00011855
CAS Number: 1779-51-7
EINECS: 217-219-4
UN: 3464
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e322e1de83c6056c1da2768b0235261af439bb0&vid=2e322e1de83c6056c1da2768b0235261af439bb0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR