Potassium citrate monohydrate

Supplier: Molekula

Synonyms: Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

21508672-1KG
MOLE21508672-1KGEA 95.3 EUR
MOLE21508672-1KG
Potassium citrate monohydrate
Potassium citrate monohydrate
Formula: HOC(COOK)(CH₂COOK)₂·H₂O
Melting Pt: 230 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 13,07706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3e6bae942870a760ea6cb418203db7458ff3dc&vid=2c2580a7bfc83e60c4dfca9f8af4ac3b3db56b27&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR