di-Ammonium oxalate monohydrate

Supplier: Molekula
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

23230264-1KG
MOLE23230264-1KGEA 172 EUR
MOLE23230264-1KG
di-Ammonium oxalate monohydrate
di-Ammonium oxalate monohydrate
Formula: (NH₄)₂C₂O₄·H₂O
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: 70 °C
Density: ∼1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 13,00541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR