1,3-Bis(diphenylphosphino)propane

Supplier: Molekula
Warning

Synonyms: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

28001437-5G
MOLE28001437-5GEA 68 EUR
MOLE28001437-5G
1,3-Bis(diphenylphosphino)propane
1,3-Bis(diphenylphosphino)propane
Formula: C₂₇H₂₆P₂
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003050
CAS Number: 6737-42-4
EINECS: 229-791-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR