2-Chloro-5-nitrobenzyl alcohol

Supplier: Molekula

Synonyms: (2-Chloro-5-nitrophenyl)methanol

29814067-5G
MOLE29814067-5GEA 0 EUR
MOLE29814067-5G
2-Chloro-5-nitrobenzyl alcohol
2-Chloro-5-nitrobenzyl alcohol
Formula: C₇H₆ClNO₃
Melting Pt: 78 °C
MDL Number: MFCD00007297
CAS Number: 80866-80-4
EINECS: 279-584-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR