G418 disulphate

Supplier: Molekula
Warning

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

M12859752
MOLEM12859752EA 0 EUR
MOLEM12859752
G418 disulphate
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR