Magnesium sulphate heptahydrate

Supplier: Molekula

Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

M20064058
MOLEM20064058EA 0 EUR
MOLEM20064058
Magnesium sulphate heptahydrate
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 2.66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR