VWR® Gentle ReView™, Stripping Buffer
Supplier: VWR Chemicals
N552-15MLEA 45.7 EUR
N552-15ML N552-500ML
VWR® Gentle ReView™, Stripping Buffer
Buffers Electrophoresis Buffers Stripping Buffers
Gentle ReView™ Stripping Buffer is a mild, yet effective method for stripping primary and secondary antibodies from PVDF or nitrocellulose membranes.
  • Odor-free, room temperature procedure
  • Minimize loss of antigen
The gentle nature of this buffer makes it possible to re-probe the same membrane several times without damaging the membrane-bound antigen. It is an odorless, ready-to-use formula that equires no mixing or heating prior to use. Most antibody-antigen complexes can be dissociated within 30 minutes at room temperature by Gentle ReView™ Stripping Buffer. However, incubation times and temperatures are dependent upon the affinity of the antibody-antigen interaction and may need to be optimized for each combination used.
Caution: For research use only. Not for therapeutic or diagnostic use.